Pierce™ Glutathione Magnetic Agarose Beads, 20 mL - FAQs
Pierce™ Glutathione Magnetic Agarose Beads, 20 mL - FAQs
View additional product information for Pierce™ Glutathione Magnetic Agarose Beads - FAQs (78602, 78601)
3 product FAQs found
Can the Pierce Magnetic Agarose Beads be frozen?
No. Freezing will adversely affect the agarose structure and may cause bead aggregation or loss of binding.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can the Pierce Magnetic Agarose Beads be used in an automated magnetic particle processor?
Yes. Protocols for using the beads on a KingFisher instrument are available on the product page.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What advantage do Pierce Magnetic Agarose beads have over Dynabeads or Pierce Magnetic Beads?
Pierce Magentic Agarose Beads have a much higher binding capacity.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.