Pierce™ Protein A/G Magnetic Agarose Beads
Thermo Scientific™

Pierce™ Protein A/G Magnetic Agarose Beads

Thermo Scientific™ Pierce™ Protein A/G Magnetic Agarose Beads provide a fast, convenient method for purification of immunoglobulins from serum, cellRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7861020 mL
786094 mL
Catalog number 78610
Price (USD)
1,763.65
Online exclusive
1,960.00
Save 196.35
Each
Quantity:
20 mL
Price (USD)
1,763.65
Online exclusive
1,960.00
Save 196.35
Each

Thermo Scientific™ Pierce™ Protein A/G Magnetic Agarose Beads provide a fast, convenient method for purification of immunoglobulins from serum, cell culture supernatant, or ascites. For antibody purification, the beads are incubated with the antibody sample and then magnetically separated from the supernatant. Nonspecifically bound serum or host cell protein can be washed away before dissociating bound antibody with elution buffer. The beads are removed from the solution manually using a magnetic stand or by automation using an instrument such as the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purification and screening of purification conditions.

Download the KingFisher™ Duo BindIt™ Software protocol (.bdz) >

Download the KingFisher™ Flex BindIt™ Software protocol (.bdz) >

Pierce Protein A/G Magnetic Agarose Beads contain a 50.5 kDa Protein A/G recombinant fusion protein that is covalently attached to a magnetite-embedded agarose core particle. These beads are not simply a mixed immobilization of separate Protein A and Protein G polypeptides, nor are they a mixture of Protein A magnetic beads and Protein G magnetic beads. The recombinant chimeric Protein A/G combines four IgG binding domains from Protein A and two binding domains from Protein G, making it a more general and convenient tool for purifying immunoglobulins.

Features of Protein A/G Magnetic Agarose beads:

  • Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables IgG purification from nearly any mammalian species
  • High capacity—sufficient for both routine and demanding purification procedures
  • Low non-specific binding—optimized purification protocol results in better IgG purification
  • Flexibility—convenience of IgG binding domains of both Protein A and Protein G on one bead
  • Compatibility— beads are compatible with manual and automated applications
  • Inert and stable—superior manufacturing method immobilizes Protein A/G by leach-resistant covalent bonds

The high-performance, magnetite-containing, superparamagnetic magnetic agarose beads are validated and optimized for use with high-throughput magnetic platforms, such as the KingFisher 96 and KingFisher Flex magnetic particle processors, but the beads also enable premium performance for simple benchtop purification applications using an appropriate magnetic stand.

Our recombinant Protein A/G binds to all human IgG subclasses, making it the ideal choice for purification of polyclonal or monoclonal IgG antibodies whose subclass identities have not been determined. In addition, it binds to IgA, IgE, IgM and (to a lesser extent) IgD. Protein A/G binds well to all mouse IgG subclasses but does not bind mouse IgA, IgM, or serum albumin. This makes Protein A/G an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses, without interference from IgA, IgM, and murine serum albumin. Individual subclasses of mouse monoclonals are likely to have a stronger affinity to the chimeric Protein A/G than to either Protein A or Protein G.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume5 mL
ConcentrationSlurry: 25%
FormatBottle
Protein FormRecombinant
Quantity20 mL
Shipping ConditionWet Ice
Stationary PhaseProtein A/G
Capacity (Metric)20 mL
FormLiquid
Particle Size25 to 30 μm
Product LinePierce
TypeMagnetic Agarose Bead
Unit SizeEach
Contents & Storage
5 mL settled resin. Supplied as 20 mL of 25% v/v suspension in phosphate buffered saline, 0.01% Tween-20 detergent, 0.05% sodium azide.

Store at 2–8°C.

Frequently asked questions (FAQs)

Can the Pierce Magnetic Agarose Beads be frozen?

No. Freezing will adversely affect the agarose structure and may cause bead aggregation or loss of binding.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can the Pierce Magnetic Agarose Beads be used in an automated magnetic particle processor?

Yes. Protocols for using the beads on a KingFisher instrument are available on the product page.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What advantage do Pierce Magnetic Agarose beads have over Dynabeads or Pierce Magnetic Beads?

Pierce Magentic Agarose Beads have a much higher binding capacity.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (4)

Citations & References
Abstract
CB1 agonism prolongs therapeutic window for hormone replacement in ovariectomized mice
Authors:Zhang KZ, Yang Q, Yang LY, Li Y, Wang X, Li Y, Dang R, Guan S, Guo Y, Sun T, Wu Y, Liu A, Zhang Y, Liu S, Zhao M
Journal:J Clin Investig
PubMed ID:
Targeting translation activity at the ribosome interface with UV-active small molecules
Authors:Kandala DT, Del Piano A, Minati L, Clamer M
Journal:ACS Omega
PubMed ID:
A vaccine targeting resistant tumours by dual T cell plus NK cell attack
Authors:Soumya Badrinath, Maxence O. Dellacherie, Aileen Li, Shiwei Zheng, Xixi Zhang, Miguel Sobral, Jason W. Pyrdol, Kathryn L. Smith, Yuheng Lu, Sabrina Haag, Hamza Ijaz, Fawn Connor-Stroud, Tsuneyasu Kaisho, Glenn Dranoff, Guo-Cheng Yuan, David J. Mooney & Kai W. Wucherpfennig
Journal:Nature
PubMed ID:
Interferon Regulatory Factors IRF1 and IRF7 Directly Regulate Gene Expression in Bats in Response to Viral Infection
Authors:Aaron T. Irving, Qian Zhang, Pui-San Kong, Katarina Luko, PritishaRozario1MingWen, Feng Zhu, PengZhou, Justin H.J. Ng, Radoslaw M. Sobota, Lin-Fa Wang
Journal:Cell Reports
PubMed ID: