Y-PER™ Yeast Protein Extraction Reagent, 500 mL - FAQs
Y-PER™ Yeast Protein Extraction Reagent, 500 mL - FAQs
View additional product information for Y-PER™ Yeast Protein Extraction Reagent - FAQs (78991, 78990)
2 product FAQs found
Can you provide the shelf-life for the Y-PER Yeast Protein Extraction Reagent?
The Y-PER Yeast Protein Extraction Reagent is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (room temperature). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Does Y-PER Yeast Protein Extraction Reagent (Cat. No. 78990) yield samples that are compatible with the Pierce BCA Protein Assay Kit (Cat. No. 23225)?
Yes, Y-PER Yeast Protein Extraction Reagent is compatible with our Pierce BCA Protein Assay Kit. Please see our Protein assay compatibility table in this Tech Tip (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.