B·R·A·H·M·S™ Free βhCG KRYPTOR™
Thermo Scientific™

B·R·A·H·M·S™ Free βhCG KRYPTOR™

B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Reach outstanding precision and lot-to-lot long term stability for trisomy screening in first and second trimester of pregnancy with B·R·A·H·M·S Free βhCG KRYPTOR.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
809075NEnsayos de inmunofluorescencia
080991NCalibrador
Número de catálogo 809075N
Precio (CLP)
-
Tipo:
Ensayos de inmunofluorescencia

B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of trisomy in first and second trimester of pregnancy. B·R·A·H·M·S Free βhCG KRYPTOR provides an early identification of the pregnancies at risk of trisomy 21, 18 and 13 in first trimester and trisomy 21 and 18 in second trimester.

US Only: Product is not 510k cleared in US.

Get confidence in your results with the high precision and the low level of interference of B·R·A·H·M·S Free βhCG KRYPTOR.

  • Save cost and time with a multiparametric control containing 3 parameters: PAPP-A, Free βhCG and AFP
  • Incubation time of only 19 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 29 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every 2 weeks
  • Ready for use reagents
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
  • Allows risk calculation in first and second trimester with B·R·A·H·M·S Fast screen pre I plus

Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Especificaciones
Tiempo de análisis19 minutes
Método de detecciónImmunofluorescence
Para utilizar con (aplicación)For quantitative determination of free beta subunit of human chrorionic gonadotropin hormone (Free βhCG). Risk assessement of fetal trisomy 21, 18 and 13 in first trimester of pregnancy, risk assessment of fetal trisomy 21 and 18 in second trimester of pregnancy
IncluyeKit for 75 determinations
Tipo de muestraSerum
TipoEnsayos de inmunofluorescencia
Analitos detectablesbeta-hCG (Free)
FormularioReady-to-Use (RTU)
Unit Size1 kit
Contenido y almacenamiento
Kit for 75 determinations, 2°C to 8°C