What is the warranty period for the Orion ROSS PerpHecT Micro Glass Bodied Combination pH Electrode?
The warranty period for the Orion ROSS PerpHecT Micro Glass Bodied Combination pH Electrode is 12 months from date of purchase.
Search
View additional product information for Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode - FAQs (8220BNWP)
1 product FAQs found