MarqMetrix™ Process BallProbe™ Sampling Optic
MarqMetrix™ Process BallProbe™ Sampling Optic
MarqMetrix™ Process BallProbe™ Sampling Optic
Thermo Scientific™

MarqMetrix™ Process BallProbe™ Sampling Optic

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is a robust immersion probe for precise, real-time compositional measurements. Its streamlined design and lack of sample preparation requirements allow users to collect reproducible spectra in seconds. Built to withstand tough conditions, it is ideal for various industries where accuracy and reproducibility are crucial. Available in multiple diameters, bundles.
製品番号(カタログ番号)プローブ長内容
840-40190010 in. (254 mm)1/2 in. Process BallProbe
840-4020008.5 in. (216 mm)3/8 in. Process BallProbe
840-40070010 in. (254 mm)Interchangeable FiberHead, 1/2 in. Process BallProbe
840-4008008.5 in. (216 mm)Interchangeable FiberHead, 3/8 in. Process BallProbe
製品番号(カタログ番号) 840-401900
価格(JPY)
-
プローブ長:
10 in. (254 mm)
内容:
1/2 in. Process BallProbe

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is an immersion probe for touch measurements in process and laboratory applications. With its streamlined design, users can easily collect reproducible spectra of liquids, solids, slurries, powders, and heterogeneous mixtures in seconds without sample preparation.

Paired with the MarqMetrix All-In-One Process Raman Analyzer, it enables precise, real-time compositional measurements. This corrosion-resistant probe features a gold compression seal and pressure design condition of 6,000 psi, making it suitable for demanding chemical, pressure, and temperature environments. It is ideal for biopharmaceutical, pharmaceutical, food and beverage processing, polymer and plastic manufacturing, and hazardous chemical applications, where measurement accuracy and reproducibility are critical.

The MarqMetrix Process BallProbe Sampling Optic – a robust immersion probe for precise, real-time compositional measurements. Its streamlined design and lack of sample preparation requirements allow users to collect reproducible spectra in seconds. Built to withstand tough conditions, it is ideal for various industries where accuracy and reproducibility are crucial. Available in multiple diameters and bundle sets.

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is an immersion probe for touch measurements in process and laboratory applications. With its streamlined design, users can easily collect reproducible spectra of liquids, solids, slurries, powders, and heterogeneous mixtures in seconds without sample preparation.

Paired with the MarqMetrix All-In-One Process Raman Analyzer, it enables precise, real-time compositional measurements. This corrosion-resistant probe features a gold compression seal and pressure design condition of 6,000 psi, making it suitable for demanding chemical, pressure, and temperature environments. It is ideal for biopharmaceutical, pharmaceutical, food and beverage processing, polymer and plastic manufacturing, and hazardous chemical applications, where measurement accuracy and reproducibility are critical.

Features and benefits

  • Immersion probe for touch measurements
  • Measures liquids, solids, slurries, powders, and heterogeneous mixtures
  • Simple design for measurement accuracy and reproducibility
  • Designed to withstand challenging chemical, pressure, and temperature conditions

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Polymer and plastic manufacturing
  • Hazardous chemical applications
  • High pressure and high temperature

Operation conditions

  • Suitable for continuous exposure to dilute and concentrated acids (hot/cold, bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
仕様
Compatible Wavelengths500 to 1100 nm
概要1/2 in. Process BallProbe Made with high purity UV-grade sapphire ball lens aligned along the C-axis, eliminating response variability due to birefringence. Avoid exposure to aqua regia.
使用対象 (装置)MarqMetrix All-In-One Process Raman Analyzer
内容1/2 in. Process BallProbe
材料Wetted Stainless Steel (C276)
光学設計6.00 mm UV-grade sapphire ball
Pressured Design Condition6,000 psi (413 bar)
プローブ長10 in. (254 mm)
Probe Outside Diameter0.5 in. (12.7 mm)
シール材質Gold
温度範囲(華氏)- 20°C to 300°C
ディスプレイNo Display
製品タイプBallProbe
Unit SizeEach