MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
Thermo Scientific™

MarqMetrix FlowCell Sampling Optic

The Thermo Scientific™ MarqMetrix™ FlowCell™ Sampling Optic is for challenging process flow-through applications. This optic ensures robust Raman measurements for your continuous flow process.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripciónConexiones
840-403000Celda de flujo de 9000 psi (dorada)1/4 in Conexiones HiP LF4
840-402700Celda de flujo de 500 psi1/8 in Racor de tubo
840-402800Celda de flujo de 2500 psi (dorada)1/8 in Racor de tubo
840-403100Celda de flujo de 4000 psi (dorada), extraíble1/8 in Prensa
840-403200Celda de flujo de 4000 psi (dorada), extraíblePrensa 1/4
840-403300Celda de flujo de 4000 psi (dorada), extraíblePrensa 3/8
840-403400Celda de flujo de 4000 psi (dorada), extraíblePrensa 1/2
840-403500Celda de flujo de 4000 psi (dorada), extraíbleAbrazadera triple
Número de catálogo 840-403000
Precio (CLP)
-
Descripción:
Celda de flujo de 9000 psi (dorada)
Conexiones:
1/4 in Conexiones HiP LF4

The Thermo Scientific™ MarqMetrix™ FlowCell™ Sampling Optic is intended for challenging process flow-through applications. The unibody design places the patented BallProbe™ spherical optic directly in the flow path, minimizing dead volume and sample handling requirements while fully utilizing optical efficiencies.

The MarqMetrix FlowCell Sampling Optic is designed for easy integration into continuous flow applications. Plumbing connections are made with standard 1/8 in. tube fitting connectors (e.g., Swagelok, Parker A-lok, Hoke, etc.). Each MarqMetrix FlowCell Sampling Optic is pressure tested so that you can quickly and confidently integrate robust Raman measurements into your continuous flow process.

Features and benefits

  • Ideal for flow-through, continuous flow applications
  • Minimizes dead volume and sample handling requirements
  • Simple design for easy integration
  • Designed to withstand challenging chemical conditions



Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Polymer and plastic manufacturing
  • Hazardous chemical applications



Operating conditions

  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
Especificaciones
Compatible Wavelengths500-1100nm
Conexiones1/4 in Conexiones HiP LF4
DescripciónCelda de flujo de 9000 psi (dorada)
Para utilizar con (equipo)MarqMetrix All-In-One Process Raman Analyzer
MaterialWetted = Inconel 625
Diseño óptico6 mm diameter UV-grade sapphire ball BallProbe
Pressured Design Condition
  • 200 μL flow path volume 
  • Individually pressure tested to 9000 psi 
Material de los cierresGold
Intervalo de temperatura (métrico)- 20°C to 350°C with gold seal
PantallaNo Display
Tipo de productoBallProbe
Unit SizeEach