Pierce™ 1-Step Transfer Buffer, 1 L - FAQs
Pierce™ 1-Step Transfer Buffer, 1 L - FAQs
View additional product information for Pierce™ 1-Step Transfer Buffer - FAQs (84731X5, 84731)
2 product FAQs found
I have the Thermo Scientific G2 Fast Blotter (Cat. No. 62288). Has this device been discontinued? Can I get the 1-Step Transfer Buffer recommended for fast transfer with the Thermo Scientific G2 Fast Blotter?
Yes. The Thermo Scientific G2 Fast Blotter has been discontinued. The direct replacement for the Thermo Scientific G2 Fast Blotter is the Thermo Scientific Power Blotter (Cat. No. 22834). The 1-Step Transfer buffer (Cat. No. 84731) is still available.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can be simultaneously transferred using the Thermo Scientific Power Blotter when used in conjunction with the Thermo Scientific 1-Step Transfer Buffer?
When the Thermo Scientific Power Blotter is used in conjunction with the Thermo Scientific 1-Step Transfer Buffer, one to four mini-sized gels or one to two midi-sized gels can be simultaneously transferred in 5-10 minutes. Please note that gels simultaneously transferred must have the same formulation.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.