Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • ELISA Kits ›
          • AKT Pan/GSK3 alpha/beta/p70 S6 Kinase ELISA Kits
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit
          Group 53 Created with Sketch.
          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit

          Representative data for AKT Pathway (AKT/GSK/p70 S6K) Activation Profile Multispecies InstantOne™ ELISA. Representative data for p38 MAPK (Total) Multispecies InstantOne™ ELISA. MCF7 cells were treated with either the PI3 Kinase inhibitor Wortmannin (10 µM) (-) or insulin (10 ng/mL for 10 min.) (+). Cell lysates were prepared as described in the InstantOne™ ELISA User Manual. Using the AKT Pathway Activa... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit
          Multispecies AKT Pathway (AKT/GSK/p70 S6K) Activation Profile InstantOne™ ELISA Kit (85-86048-11)
          videoThumbnail
          ►

          Product Specifications

          Sample type/volume

          Cell lysate
          20 µL

          Hands-on time

          1 hr 20 min

          Time-to-result

          1 hr 10 min

          Homogenous (no wash)

          No

          Instrument

          Absorbance Microplate Reader

          Product size

          96 Tests

          Contents

          Capture Antibody
          Detection Antibody Reagent
          InstantOne™ Assay Plate
          Cell Lysis Buffer
          Enhancer Solution
          Wash Buffer (10X)
          Substrate Reagent (TMB)
          Stop Solution
          Positive Control Cell Lysate (lyophilized)
          Plate seal

          Shipping conditions

          Wet ice

          Storage

          2-8°C

          Protein name

          AKT Pan/GSK3 alpha/beta/p70 S6 Kinase

          Target site-specificity

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated AKT 1/2/3 (Ser473) (human, mouse, hamster, rat), phosphorylated GSK3beta (Ser9) (human, mouse, rat) and phosphorylated p70S6K (Thr389) (human, mouse) in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

          Target information

          AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). Activation of PI3K results in the generation of phosphatidylinositol (3,4,5)-triphosphate (PIP3). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. AKT phosphorylates over 50 known substrates, including GSK3b, p70S6K, AS160, PRAS40, TSC 1, TCS 2, Raf-1, Bad, the FOXO family of transcription factors, and PFK2. Due to its established critical role in numerous biological processes, AKT is a frequent target for oncology-related drug development. Glycogen Synthase Kinase 3 (GSK3) is a Ser/Thr kinase that exists as two isoforms, GSK3a and GSK3β. Unlike other kinases, phosphorylation at Ser9 and Ser21 for GSK3alpha and GSK3beta, respectively, leads to inactivation of this enzyme. GSK3 has been implicated in several diseases, including cancer and diabetes, through its phosphorylation of glycogen synthase. GSK3 has also been associated with neurodegenerative diseases such as Alzheimer's disease due to its phosphorylation of the neuronal Tau protein. p70 S6 Kinase (p70S6K) is a member of the ribosomal S6 kinase family of Ser/Thr kinases. P70 S6K activity is controlled by multiple signaling pathways, including the MAPK, phosphoinositide-3 kinase (PI3K), and mTOR pathways. This regulation results in the phosphorylation of multiple sites of p70 S6K, including Thr389 within the linker domain, which is required for full activation. Activated p70 S6K phosphorylates several residues on the S6 ribosomal protein, thereby leading to increased protein synthesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0