Tween™ 20 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Tween™ 20 Surfact-Amps™ Detergent Solution

Thermo Scientific Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween™ 20 detergent stabilized as a 10% solution (w/v) and packagedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8511350 mL
283206 x 10 mL
85114250 mL
85115500 mL
283211 L
Catalog number 85113
Price (USD)
168.65
Online exclusive
185.00
Save 16.35
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
168.65
Online exclusive
185.00
Save 16.35
Each
Thermo Scientific Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween™ 20 detergent stabilized as a 10% solution (w/v) and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring it's stability and eliminating the accumulation of peroxides and degradation products.

Features of Tween-20 Surfact-Amps Detergent Solution:

• Tween 20—a popular nonionic detergent for use in ELISA, Western blotting and other immunoassay PBS or TBS wash buffers
• Accurate—precise 10% detergent solution in ultrapure water
• Easy-to-use—solution is simple to dispense and dilute for use
• Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
• Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Properties of Tween 20:
• Molecular Weight: 1228g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: Unknown
• Micelle Molecular Weight: Unknown
• Critical Micelle Concentration (CMC): 0.06 mM (0.0074%, w/v)
• Cloud Point: 95°C
• Dialyzable: No

Specifications for Tween 20 Surfact-Amps Detergent Solution:
• Visual: Clear, light yellow liquid, free of particulates.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity50 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineSurfact-Amps, Tween
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in a cool, dry, and well-ventilated area

Frequently asked questions (FAQs)

What is the Thermo Scientific Tween-20 Surfact-Amps Detergent Solution?

Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween 20 detergent stabilized as a 10% solution and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring its stability and eliminating the accumulation of peroxides and degradation products.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What quality testing is performed for your Tween 20 Surfact-Amps Detergent Solution?

The detergent is screened for:
- Visualization: clear, light yellow liquid, free of particulates
- Concentration: 10.0 +/- 1.0%
- Oxidant concentration: less than or equal to 1.0 µeq/mL
- Carbonyl concentration: less than or equal to 1.0 µeq/mL
- suspended solids: residue present must not exceed Residue Reference

See the COA for further details.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the main advantages of using Thermo Scientific Pierce Surfact-Amps detergents?

Thermo Scientific Pierce Surfact-Amps detergents are highly purified, precisely diluted (10%) formulations that are ideal for applications or assays that are sensitive to contaminants that are present in unpurified detergents. We test every batch to insure that our detergents contain less than 1.0 µeq/mL peroxides and carbonyls and package them under nitrogen, to prevent oxidization during storage.

Superior quality - lower measurable contaminant levels than other leading vendors
Accurate -precise 10% detergent solution in ultrapure water
Easy-to-use - solution is simple to dispense and dilute for use
Exceptionally pure - less than 1.0 µeq/mL peroxides and carbonyls

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Are detergents denaturing or non-denaturing with respect to protein structure?

Ionic detergents, or those that carry a charge, are the most likely to be denaturing to proteins. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking protein-protein interactions through changes in the three-dimensional structure of the proteins. Nondenaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS).



Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What are detergents?

Detergents are amphipathic molecules containing both a nonpolar “tail” having aliphatic or aromatic character, and a polar “head”. The ionic character of the polar head group forms the basis for broad classification of detergents as ionic, nonionic, or zwitterionic.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.