Pierce™ Tissue Strainers, 250 μm, 2.5 mL - FAQs
Pierce™ Tissue Strainers, 250 μm, 2.5 mL - FAQs
View additional product information for Pierce™ Tissue Strainers, 250 μm, 2.5 mL - FAQs (87791)
1 product FAQs found
Can I autoclave Pierce Tissue Strainers, 250 µm, 2.5 mL (Cat. No. 87791)?
Pierce Tissue Strainers, 250 µm, 2.5 mL (Cat. No. 87791), can be autoclaved at 121 degrees C for 20-30 min without loss of function. Because they are made from nylon, they cannot withstand higher temperatures or longer durations.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.