eBioscience™ Annexin V Apoptosis Detection Set PE-Cyanine7, 50 Tests - FAQs
eBioscience™ Annexin V Apoptosis Detection Set PE-Cyanine7, 50 Tests - FAQs
View additional product information for eBioscience™ Annexin V Apoptosis Detection Kits - FAQs (88-8005-74, BMS500BT-100, 88-8103-72, BMS500BT-300, BMS500FI-20, 88-8102-72, BMS500BT-20, BMS500FI-100, 88-8103-74, 88-8102-74, BMS500FI-300, 88-8008-72, 88-8006-72, 88-8007-72, 88-8006-74, 88-8005-72, 88-8007-74, 88-8008-74)
5 product FAQs found
Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?
Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.
I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?
Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).
Can I detect annexin V staining in an imaging assay?
Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.
When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?
Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).
Can I fix my cells after annexin V staining?
Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.