Product Image
Invitrogen™

eBioscience™ Mouse Regulatory T Cell Staining Kit #1

The FJK-16s antibody reacts with mouse/rat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of thisRead more
Have Questions?
Catalog NumberQuantity
88-8111-401 kit
Catalog number 88-8111-40
Price (USD)
611.65
Online exclusive
816.00
Save 204.35
Each
Quantity:
1 kit
Price (USD)
611.65
Online exclusive
816.00
Save 204.35
Each
The FJK-16s antibody reacts with mouse/rat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3 a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.

Intracellular staining of mouse splenocytes with FJK-16s using the mouse Foxp3 staining sets and protocol reveals approximately 2% of total splenocytes in the C57Bl/6 strain and approximately 3-5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+CD25+ cells and 4% of the CD4+CD25- cells staining with FJK-16s. B220+, CD11b+, CD11c+, and Ly6G/Gr-1+ cells do not show significant co-staining with FJK-16s. These data are consistent with a recent report which follows expression of Foxp3 using a GFP knock-in (Fontenot et al, 2005).

FJK-16s cross-reacts with rat Foxp3 This has been demonstrated by intracellular staining of Foxp3 and flow cytometry of rat splenocytes using the same method and reagents as used for mouse tissue. Please note that the CD4 and CD25 antibodies included in this kit only recognize the mouse antigens. For staining rat tissue, please use (CD4 FITC cat. 11-0040 and CD25 APC cat. 17-0390)

The anti-mouse Foxp3 Staining Set has been formulated and optimized for the staining of mouse splenocytes with the FJK-16s monoclonal antibody

Not included:
Isotype controls for anti-CD4 (rat IgG2a, cat. 11-4321) and anti-CD25 (rat IgG1, cat. 17-4301)

Host
Rat

Isotype
IgG2a

Reactivity/Species
Mouse

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.2mg/mL
Quantity1 kit
For Use With (Application)Intracellular Staining, Flow Cytometry
Product LineeBioscience
Product TypeCell Staining Kit
Unit SizeEach
Contents & Storage
2° to 8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.

Frequently asked questions (FAQs)

I want to obtain good results, so which format of Ebioscience Foxp3 antibody should I order?

Our recommendation is to use the Foxp3 antibodies in PE or APC as they are brighter than FITC. Furthermore we prefer CD4 in FITC since the expression levels are high enough to use with this fluorochrome. We then use CD25 in APC. Please see our Treg staining Kits for prebundled kits with optimal fluorochrome combinations (Cat. No. 88-8999-40 and 88-8111-40).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.