Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • IL-17AF ELISA Kits
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          Mouse IL-17AF (heterodimer) Uncoated ELISA Kit with Plates

          1 Reference

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Mouse IL-17AF (heterodimer) Uncoated ELISA Kit with Plates (88-8711-76)
          Group 53 Created with Sketch.
          Mouse IL-17AF (heterodimer) Uncoated ELISA Kit with Plates (88-8711-76)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Mouse IL-17AF (heterodimer) Uncoated ELISA Kit with Plates

          Uncoated ELISA Kit with Plates {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Mouse IL-17AF (heterodimer) Uncoated ELISA Kit with Plates (88-8711-76)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          16 pg/mL

          Assay range

          16-2,000 pg/mL

          Sample type/volume

          Serum
          50 µL
          Supernatant
          50 µL

          Hands-on time

          1 hr 45 min

          Time-to-result

          24 hr 30 min

          Homogenous (no wash)

          No

          Instrument

          Absorbance Microplate Reader

          Product size

          20 x 96 Tests

          Contents

          Capture Antibody: Pre-titrated, purified antibody
          Detection Antibody: Pre-titrated, HRP-conjugated antibody
          Standard: Recombinant protein for generating standard curve and calibrating samples
          10X Coating Buffer: Buffer for plating the Capture Antibody
          5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme
          Substrate: 1X TMB Solution
          Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
          96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)

          Storage

          2-8°C

          Protein name

          IL-17AF

          Target site-specificity

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          The Mouse Interleukin 17AF (heterodimer) (IL-17AF) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17AF(heterodimer). Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately.

          Principle of the method

          ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

          Target information

          IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases : IL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8, IL-17F

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch