I want to obtain good results, so which format of Ebioscience Foxp3 antibody should I order?
Our recommendation is to use the Foxp3 antibodies in PE or APC as they are brighter than FITC. Furthermore we prefer CD4 in FITC since the expression levels are high enough to use with this fluorochrome. We then use CD25 in APC. Please see our Treg staining Kits for prebundled kits with optimal fluorochrome combinations (Cat. No. 88-8999-40 and 88-8111-40).