MagniSort™ Human CD4 Naïve T cell Enrichment Kit - FAQs
MagniSort™ Human CD4 Naïve T cell Enrichment Kit - FAQs
View additional product information for MagniSort™ Human CD4 Naïve T cell Enrichment Kit - FAQs (8804-6814-74)
1 product FAQs found
I need to further purify my naive human T cells that have been isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit (Cat. No. 8804-6814-74). Which purification kit can I use?
The CD4 naive T cells that are isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit are untouched and free in solution. Hence, you can use any other purification kit afterwards.