HisPur™ Ni-NTA Resin
Thermo Scientific™

HisPur™ Ni-NTA Resin

Thermo Scientific HisPur Ni-NTA Resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.Features ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8822110 mL
88222100 mL
88223500 mL
Catalog number 88221
Price (USD)
134.65
Online exclusive
158.00
Save 23.35
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
134.65
Online exclusive
158.00
Save 23.35
Each
Thermo Scientific HisPur Ni-NTA Resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.

Features of HisPur Ni-NTA Resin:

• High capacity—bind up to 60 mg of 6xHis-tagged protein per milliliter of resin
• Versatile—purify proteins using native or denaturing conditions
• Compatible—use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
• Cost-effective—reuse the same batch of resin at least five times
• Easy to use—pre-formulated buffers available for kit formats
• Flexible—available in multiple formats including bulk resin, spin columns, chromatography cartridges and 96-well filter plates

The specially prepared support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

The expression and purification of recombinant proteins is central to protein regulation, structure and function studies. The majority of recombinant proteins are expressed as fusions with short affinity tags, the most popular being the polyhistidine (6xHis) tag. The method used to purify recombinant His-tagged proteins is immobilized metal affinity chromatography (IMAC), consisting of chelating resins charged with either nickel or cobalt ions that coordinate with the histidine side chains.

HisPur Ni-NTA Resin effectively purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates, such as those that are attained with Thermo Scientific B-PER Bacterial Protein Extraction Reagents. The resin performs well in batch-binding and spin-column procedures at a variety of scales. Performance equals or exceeds popular Ni-NTA resins from other suppliers. HisPur Ni-NTA Resin is a high-quality, stable and resilient affinity support. Tests confirm that no decrease in performance occurs after at least five repeated uses. These data indicated that the resin is highly resistant to structural degradation or nickel ion leaching during normal use.

HisPur Ni-NTA Resin and HisPur Cobalt Resin are alternative forms of IMAC, the former using nickel and the latter using cobalt as the chelated metal ion responsible for His-tag binding. Ni-NTA resins have been the most common IMAC resin choice for 6xHis-tag protein purifications because of the four metal-binding sites on the chelate, which enables high-protein binding and low-metal ion leaching. Cobalt binds less strongly and therefore more discriminantly, enabling greater purity but usually less yield in His-tagged protein purification.

Available Formats:
• Resin Slurries—crosslinked 6% beaded agarose; 10 mL, 100 mL, 500 mL bottles
• Spin Columns—0.2 mL (microcentrifuge), 1 mL, 3 mL columns
• Purification Kits—complete kits in all three column sizes
• Chromatography Cartridges—learn more and see HisPur Chromatography Cartridge data
• 96-well Spin Plates

Related Products
HisPur™ Ni-NTA Spin Columns, 0.2 mL
HisPur™ Ni-NTA Spin Columns, 1 mL
HisPur™ Ni-NTA Spin Columns, 3 mL
HisPur™ Ni-NTA Chromatography Cartridges, 1 mL
HisPur™ Ni-NTA Superflow Agarose
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, Agarose Resin
DescriptionHisPur Ni-NTA Resin
FormatBottle
Quantity10 mL
Stationary PhaseDivalent Nickel, Metal Chelate
TargetHis-Tagged Protein
Binding CapacityUp to 60 mg of 6xHis-Tagged Protein Binding/mL Resin
FormLiquid Suspension
Product LineHisPur
TypeResin
Unit SizeEach
Contents & Storage
Store at 4°C

Frequently asked questions (FAQs)

Are there other sequences that can bind to nickel more tightly than 6xHis-tagged proteins and how can they be eluted.

Yes, 7xHis-tagged proteins, proteins naturally high in histidine, and other combinations of His and other amino acids will bind. To elute them, you have to increase the concentration of imidazole. Generally these peptides will not contaminate your fraction since they remain on the column. However after multiple uses of the same column, these peptides may reduce the binding capacity of the column.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can the HisPur Ni-NTA Spin plates (Cat. No. 88230) be regenerated and reused, or are they for single-use only?

The resin can be re-used at least five times; this is also for the resin in the plates. Regeneration is possible using this procedure (https://assets.thermofisher.com/TFS-Assets%2FLSG%2Fmanuals%2FMAN0011700_HisPur_NiNTA_Resin_UG.pdf) Since the resin bed volume in each well is 50 µL, you can use 250 µL twice to give a total of 10 resin-bed volume.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How do I perform Ni-NTA bead regeneration with the HisPur Ni-NTA Resin (Cat. No. 88221)?

Please refer to page 2 (buffer preparation) and page 3 (procedure) of the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011700_HisPur_NiNTA_Resin_UG.pdf) for information regarding bead regeneration.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can ProBond or Ni-NTA beads be used for large-scale preparations?

ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?

Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.