SnakeSkin™ Dialysis Tubing, 10K MWCO, 16 mm, 10.5 m - FAQs
SnakeSkin™ Dialysis Tubing, 10K MWCO, 16 mm, 10.5 m - FAQs
View additional product information for SnakeSkin™ Dialysis Tubing - FAQs (68035, 68700, 68100, 88245, 88244, 68011, 88243, 88242)
2 product FAQs found
What is the difference between your dialysis devices (plates, device, cassettes, flasks)?
Please view our selection table to choose the right dialysis device for your experiment - https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-dialysis-desalting-concentration/dialysis-products.html.
What is protein dialysis?
Dialysis is the separation of small and large molecules in a solution by selective diffusion through a semi-permeable membrane. It is generally used for larger sample volumes, and can take hours to overnight for complete dialysis.