Pierce™ D-Luciferin, Monopotassium Salt
Actual product may vary
Thermo Scientific™

Pierce™ D-Luciferin, Monopotassium Salt

Thermo Scientific Pierce D-Luciferin, Monopotassium Salt is formulated at greater than 99% purity to provide convenience and high performance atRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
882941 g
88293100 mg
Catalog number 88294
Price (USD)
682.65
Online exclusive
750.00
Save 67.35
Each
Quantity:
1 g
Price (USD)
682.65
Online exclusive
750.00
Save 67.35
Each

Thermo Scientific Pierce D-Luciferin, Monopotassium Salt is formulated at greater than 99% purity to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Features of D-Luciferin, Monopotassium Salt:

  • Molecular mass = 318.42 g/molHigh purity, typically greater than 99.7%, as verified by HPLC and FTIR
  • Great performance with in vitro applications
  • Suitable for in vivo imaging

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium salt of firefly luciferin is stable and easily soluble, and is compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Applications:

  • Gene reporter assays
  • ATP assays
  • High throughput sequencing
  • Contamination assays
  • Bioluminescent in vivo imaging
  • Drug screening
  • Multiplexed assays to study off-target effects
  • Signal transduction pathway analysis

Related Products

Pierce™ D-Luciferin, Monosodium Salt (Cat. No. 88291)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ATP Assay, Gene Reporter Assay
Product LinePierce
Product TypeD-Luciferin, Monopotassium Salt
Purity>99%
Quantity1 g
Salt FormMonopotassium Salt
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
Detection MethodBioluminescence
Unit SizeEach
Contents & Storage
Store at or below -20°C.