1-Step Heavy Protein IVT Kit - FAQs
1-Step Heavy Protein IVT Kit - FAQs
View additional product information for 1-Step Heavy Protein IVT Kit - FAQs (88330)
1 product FAQs found
Do you offer an alternative product to the discontinued 1-Step Heavy Protein IVT Kit (Cat. No. 88330) to enable synthesis of heavy-labelled proteins of interest?
We do not offer any alternatives for 1-Step Heavy Protein IVT Kit.