Pierce™ 6 Protein Digest, equimolar, LC-MS grade - FAQs
Pierce™ 6 Protein Digest, equimolar, LC-MS grade - FAQs
View additional product information for Pierce™ 6 Protein Digest, equimolar, LC-MS grade - FAQs (88342)
5 product FAQs found
What are the UniProt IDs for the proteins in the Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342) standard?
The UniProt IDs for the proteins in the Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342) standard, are:
    Serum Albumin, Bovine (Uniprot: P02769)
    Cytochrome C, Bovine (Uniprot: P62894)
    Lysozyme, Chicken (Uniprot: P00698)
     β-galactosidase, E. coli (Uniprot: P00722)
    Apo-Transferrin, Bovine (Uniprot: Q29443)
    Alcohol Dehydrogenase, S. cerevisiae (Uniprot: P00330)
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center or Mass Spectrometry Support Center.
How do I verify the LC-MS system after installation of a new column for proteomic applications?
Several technical replicate injections of a known standard sample, e.g., Pierce BSA Digest (Cat. No. 88341), Pierce 6 Protein Digest (Cat. No. 88342), or Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) should be used to equilibrate and compare the new column to reference chromatograms.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How do I assess my LC-MS system performance?
The best way to assess the performance of an entire LC-MS system is using a mass spectrometry calibration solution, peptide retention time standards (Pierce Peptide Retention Time Calibration Mixture (PRTC), Cat. No. 88321 and Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix), Cat. No. A40010), and a known standard sample, e.g., Pierce BSA Digest (Cat. No. 88341), Pierce 6 Protein Digest (Cat. No. 88342), or Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) specific to your application. We recommend frequent assessment of LC-MS system performance on a routine basis to compare results over time.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
I don't need a complex digest for my MALDI study, but I need something more than a single protein digest. What do you recommend?
We recommend using the moderately complex Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Which standards and calibrants do you recommend using for assessing LC-MS system suitability?
Here are our recommendations:
- Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321) for LC system and gradient optimization
- Pierce BSA Protein Digest, MS grade (Cat. No. 88341), Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342), and Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) for "bottom up" LC-MS method optimization and LC-MS suitability assessment
- Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) (Cat. No. A40010) for assessing the gradient, sensitivity, and dynamic range of the LC-MS system for discovery and targeted peptide quantitative workflows
- Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for assessing system performance for TMT-based quantitative workflows
- Pierce Intact Protein Standard Mix (Cat. Nos. A33526, A33527) for "top down" LC-MS method optimization and LC-MS suitability assessment
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.