MEM for SILAC
Thermo Scientific™

MEM for SILAC

Thermo Scientific™ MEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC)Read more
Have Questions?
Catalog NumberQuantity
88368500 mL
Catalog number 88368
Price (USD)
82.88
Special offer
Online exclusive
Ends: 15-Jul-2026
97.50
Save 14.62 (15%)
Each
Quantity:
500 mL
Price (USD)
82.88
Special offer
Online exclusive
Ends: 15-Jul-2026
97.50
Save 14.62 (15%)
Each
Thermo Scientific™ MEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of MEM for SILAC:
• Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
• High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products
NeuCode™ Lysine -080, 50 mg
Fetal Bovine Serum, dialyzed, US origin
DMEM for SILAC

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLiquid Medium
For Use With (Equipment)Mass Spectrometer
Label or DyeMass Isotope of Amino Acid
Quantity500 mL
Shipping ConditionRoom Temperature
Workflow StepCell Culture
Detection MethodMass Spectrometry
FormLiquid
Product TypeMedia
Starting MaterialProtein Samples
Unit SizeEach
Contents & Storage
1 x 500 mL, store at 2–8°C