Pierce™ NHS-Activated Magnetic Beads
Thermo Scientific™

Pierce™ NHS-Activated Magnetic Beads

Immobilize proteins or other ligands for magnetic separation affinity purification with these magnetic particles coated with amine-reactive NHS-ester groups.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
888275 mL
888261 mL
Catalog number 88827
Price (USD)
1,125.65
Online exclusive
1,224.00
Save 98.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
1,125.65
Online exclusive
1,224.00
Save 98.35
Each

Thermo Scientific Pierce NHS-Activated Magnetic Beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.

Features of NHS-Activated Magnetic Beads:

  • High capacity—at least four times higher binding capacity than NHS-activated magnetic beads from other suppliers
  • Easy to use—immobilize in a simple one-step reaction with minimal hands-on time
  • Safe—no hazardous chemicals needed (e.g., sodium cyanoborohydride and cyanogen bromide)
  • Ligand-compatible—use to immobilize with nearly any primary amine-containing compound or affinity ligand
  • Low non-specific binding—the bead surface is pre-blocked and any nonreacted NHS-ester groups are fully quenched
  • Protocol-compatible—protein coupling to the beads and downstream applications can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)

The activated magnetic beads contain N-hydroxy-succinimide (NHS) functional groups that react with primary amines forming stable amide linkages. Once they are covalently attached, the immobilized proteins are highly resistant to leaching from the bead surface. When prepared beads are used in experiments, nonspecific binding is negligible because nonreacted NHS-ester groups are thoroughly blocked during the coupling procedure. Pierce NHS-Activated Magnetic beads can be coupled and processed either manually with a magnetic stand or with automated platforms such as the Thermo Scientific KingFisher Instruments.

Applications:

  • Immobilization of ligands for the purification of recombinant proteins
  • Immobilization of antibodies for immunoprecipitations and co-immunoprecipitations free of antibody contamination in the eluates
  • Immobilization of secondary antibody for the affinity purification of antibody subtypes from ascites, serum and cell culture supernatant

Pierce NHS-Activated Magnetic Beads offer a convenient way to conjugate any desired protein to a magnetic bead surface. The process does not require hazardous chemicals or lengthy reaction schemes. The beads are first incubated with protein for 1 to 2 hours in an amine free buffer at pH 7 to 9 to allow for covalent coupling through NHS-ester chemistry. The beads are subsequently washed and then any remaining active NHS-ester groups are quenched. The amine reactive chemistry and subsequent quench are completed in 3 to 4 hours. Coupled protein does not leach from the bead surface. In addition, the beads exhibit very low non-specific binding due to effective quenching and a proprietary blocking agent that coats the surface of the base particle. Following conjugation the prepared magnetic beads are typically used in affinity purification procedures.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Pierce NHS-Activated Magnetic Beads offer a convenient way to conjugate any desired protein to a magnetic bead surface. The process does not require hazardous chemicals or lengthy reaction schemes. The beads are first incubated with protein for 1 to 2 hours in an amine free buffer at pH 7 to 9 to allow for covalent coupling through NHS-ester chemistry. The beads are subsequently washed and then any remaining active NHS-ester groups are quenched. The amine reactive chemistry and subsequent quench are completed in 3 to 4 hours. Coupled protein does not leach from the bead surface. In addition, the beads exhibit very low non-specific binding due to effective quenching and a proprietary blocking agent that coats the surface of the base particle. Following conjugation the prepared magnetic beads are typically used in affinity purification procedures.
Specifications
ConcentrationSlurry: 10 mg/mL, 1% solids
Protein FormRecombinant
Quantity5 mL
Surface FunctionalityNHS Ester
TargetAmine
Capacity (Metric)5 mL
FormLiquid
Particle Size1 μm
Product LinePierce
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product shipped with an ice pack.

Frequently asked questions (FAQs)

Can you provide the shelf-life for NHS-Activated Magnetic Beads?

NHS-Activated Magnetic Beads is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What should I do to increase the coupling efficiency of antibodies to the Pierce NHS-Activated Magnetic Beads?

-Make sure all primary amine-containing buffer components of the buffer like Tris or glycine are completely removed by dialysis, desalting, or the use of the Pierce Antibody Clean-Up Kit (Cat. No. 44600) before coupling to magnetic beads
-Make sure the antibody is mixed with the NHS-beads IMMEDIATELY after washing the beads. A delay may cause hydrolysis of the NHS groups on the resin which leads to low coupling efficiency.
-Finally make sure the pH of the non-amine-containing coupling buffer used for coupling is between pH 7-9

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.