Pierce™ Anti-HA Magnetic Beads
Thermo Scientific™

Pierce™ Anti-HA Magnetic Beads

Perform immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
888375 mL
888361 mL
Catalog number 88837
Price (USD)
1,123.65
Online exclusive
1,322.00
Save 198.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
1,123.65
Online exclusive
1,322.00
Save 198.35
Each

Thermo Scientific Pierce Anti-HA Magnetic Beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Features of Anti-HA Magnetic Beads:

  • Specific—highly specific anti-HA monoclonal antibody (clone 2-2.2.14) enables high yield and high purity immunoprecipitation
  • Convenient and fast—product instructions provide an easy-to-follow, optimized protocol for immunoprecipitation in approximately one hour
  • Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding for HA-tag IP or co-IP experiments
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is coated with anti-HA antibody, a highly specific mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein. The Pierce Anti-HA Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Specifications:

Product Details:

Pierce Anti-HA Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant HA-tagged proteins and the co-immunoprecipitation (co-IP) of their interacting proteins. The beads are incubated with a cell lysate containing HA-tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine, pH 2.0, 50 mM NaOH, or SDS-PAGE sample buffer. If gentler elution conditions are desired, 2 mg/mL Pierce HA peptide can be used. The protocol has been optimized for each of these conditions. Anti-HA antibody can be used to detect HA-tagged protein by Western blot analysis.

For Research Use Only. Not for use in diagnostic procedures.
Thermo Scientific™ Pierce Anti-HA Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant HA–tagged proteins and the co–immunoprecipitation (co–IP) of their interacting proteins. The beads are incubated with a cell lysate containing HA–tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine, pH 2.0, 50mM NaOH, or SDS–PAGE sample buffer. If gentler elution conditions are desired, 2mg/mL Pierce HA peptide can be used. The protocol has been optimized for each of these conditions. Anti-HA antibody can be used to detect HA–tagged protein by Western blot analysis.
Specifications
ConcentrationSlurry: 10mg/mL, 1% solids
Ligand TypeAntibody
Protein FormRecombinant
Quantity5 mL
TargetHA
Capacity (Metric)5 mL
FormLiquid
Particle Size1 μm
Product LinePierce
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Store at 4°C.

Frequently asked questions (FAQs)

Are the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) compatible with sodium dodecyl sulfate? If yes, up to which concentration?

Yes, the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) are compatible with sodium dodecyl sulfate, but the concentration should not exceed 0.5%.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What type of recombinant protein purification resins do you offer?

We offer a variety of resins targeting His-tagged fusion proteins or GST fusion proteins. Please see this selection table to choose the right resin for your experiments. Table available at http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/recombinant-protein-purification.html

We also offer resins (both agarose and magnetic bead-based) with immobilized anti-c-Myc and anti-HA antibodies for targeting c-Myc and HA-tagged proteins respectively. Please check out the following links for the specific products:

-Anti-c-Myc agarose (https://www.thermofisher.com/order/catalog/product/20169?ICID=search-product)
-Anti-c Myc magnetic beads (https://www.thermofisher.com/order/catalog/product/88843?ICID=search-product)
-Anti-HA agarose (https://www.thermofisher.com/order/genome-database/antibody/HA-Epitope-Tag-Antibody-clone-2-2-2-14-Monoclonal/26181)
-Anti-HA magnetic beads: (https://www.thermofisher.com/order/catalog/product/88837?ICID=search-product)


Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.