Pierce™ Anti-HA Magnetic Beads, 5 mL - FAQs
Pierce™ Anti-HA Magnetic Beads, 5 mL - FAQs
View additional product information for Pierce™ Anti-HA Magnetic Beads - FAQs (88837, 88836)
2 product FAQs found
Are the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) compatible with sodium dodecyl sulfate? If yes, up to which concentration?
Yes, the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) are compatible with sodium dodecyl sulfate, but the concentration should not exceed 0.5%.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What type of recombinant protein purification resins do you offer?
We offer a variety of resins targeting His-tagged fusion proteins or GST fusion proteins. Please see this selection table to choose the right resin for your experiments. Table available at http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/recombinant-protein-purification.html
We also offer resins (both agarose and magnetic bead-based) with immobilized anti-c-Myc and anti-HA antibodies for targeting c-Myc and HA-tagged proteins respectively. Please check out the following links for the specific products:
-Anti-c-Myc agarose (https://www.thermofisher.com/order/catalog/product/20169?ICID=search-product)
-Anti-c Myc magnetic beads (https://www.thermofisher.com/order/catalog/product/88843?ICID=search-product)
-Anti-HA agarose (https://www.thermofisher.com/order/genome-database/antibody/HA-Epitope-Tag-Antibody-clone-2-2-2-14-Monoclonal/26181)
-Anti-HA magnetic beads: (https://www.thermofisher.com/order/catalog/product/88837?ICID=search-product)
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.