Pierce™ Protein A Magnetic Beads
Thermo Scientific™

Pierce™ Protein A Magnetic Beads

Perform antibody purification and immunoprecipitation methods using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
888451 mL
888465 mL
Catalog number 88845
Price (USD)
206.65
Online exclusive
232.00
Save 25.35
Each
Quantity:
1 mL
Request bulk or custom format
Price (USD)
206.65
Online exclusive
232.00
Save 25.35
Each

Thermo Scientific Pierce Protein A Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

Features of Protein A Magnetic Beads:

  • High efficiency – equivalent or higher yield of IP target antigens than magnetic beads from other suppliers
  • Low non-specific binding – stable, pre-blocked beads provide highly purified product (e.g., antigen eluted in IP with antibody is devoid of contaminating protein from complex cell lysate)
  • Consistent – magnetic beads eliminate resin loss and provide for more efficient separation than traditional IP methods that use only centrifugation
  • Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

Recombinant protein A, covalently attached to a blocked magnetic bead surface, can bind to antibodies from many different species, enabling purification of antibodies from crude extracts. Immunoprecipitation assays performed with protein A coated beads result in high yield of target antigen with very low background. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in IP or Co-IP experiments. The Pierce Protein A Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Applications:

  • IP and Co-IP experiments
  • Antibody purification

Thermo Scientific Pierce Protein A Magnetic Beads are used for purifying antibody from serum, cell culture supernatant or ascites, as well as for IP/Co-IP of antigens from cell or tissue extracts. Protein A can bind to antibodies from many different species, including mouse, human, rabbit, pig, dog, and cat. The protocol for the Pierce Protein A beads has been optimized to allow for high recovery and high purity of the isolated antibody or antigen. Antibody or antigen/antibody complex (IP) is first captured on the magnetic beads. The beads are washed and then the target is eluted with low pH elution buffer. IP performance is equivalent to or better than Protein A magnetic beads from other suppliers.

For Research Use Only. Not for use in diagnostic procedures.
Thermo Scientific™ Pierce Protein A Magnetic Beads are used for purifying antibody from serum, cell culture supernatant or ascites, as well as for IP/Co–IP of antigens from cell or tissue extracts. Protein A can bind to antibodies from many different species, including mouse, human, rabbit, pig, dog, and cat. The protocol for the Pierce Protein A beads has been optimized to allow for high recovery and high purity of the isolated antibody or antigen. Antibody or antigen/antibody complex (IP) is first captured on the magnetic beads. The beads are washed and then the target is eluted with low pH elution buffer. IP performance is equivalent to or better than Protein A magnetic beads from other suppliers.
Specifications
Capacity (Metric)1 mL
ConcentrationSlurry: 10mg/mL, 1% solids
Ligand TypeProtein A
Protein FormRecombinant
Quantity1 mL
TargetAntibody
FormLiquid
Particle Size1 μm
Product LinePierce
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product shipped with an ice pack.

Frequently asked questions (FAQs)

We would like to crosslink our antibodies to Protein A magnetic beads rather than switch to the Protein A/G beads. Will the protocol and reagents described in the protocol for the Protein A/G beads work for the Protein A beads?

Protein A as well as Protein A/G bind to antibodies via the Fc portion. You can use DSS crosslinker to crosslink the antibody to Protein A as we described it for Protein A/G in our kit (Cat No. 88805). Please note that with this chemical crosslinking, there is the potential to lose antigen binding sites, and thus, the IP yield may be a less than when performing the IP without crosslinking.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What are the differences between Protein A, Protein G, Protein A/G, and Protein L Ig-binding proteins?

Protein A, Protein G, and Protein A/G bind almost exclusively to the IgG class of antibodies, but their binding properties differ among species and subclasses of IgG. Protein L binds in the variable fragment of some kappa light chains and can react with any immunoglobulin, not just IgG, as long as the correct kappa light chains are present. Protein L does not bind lambda light chains and certain kappa chains of different species.

-Protein A is generally preferred for rabbit, pig, dog, and cat IgG.
-Protein G has better binding capacity for a broader range of mouse and human IgG subclasses (e.g., IgG1 vs. IgG2)
-Protein A/G is a recombinant fusion protein that includes the IgG-binding domains of both Protein A and Protein G. Therefore, Protein A/G is ideal for binding the broadest range of IgG subclasses from rabbit, mouse, human, and other mammalian samples.
-Protein L binds to certain immunoglobulin kappa light chains. Because kappa light chains occur in members of all classes of immunoglobulin (i.e., IgG, IgM, IgA, IgE and IgD), Protein L can purify these different classes of antibody. However, only those antibodies within each class that possess the appropriate kappa light chains will bind. Generally, empirical testing is required to determine if Protein L is effective for purifying a particular antibody. It binds only Vk1 in mouse and VkI, VkIII and VkIV in human.
Read more about the general characteristics of Ig-binding proteins (https://www.thermofisher.com/us/en/home/life-science/antibodies/antibody-purification-kits-reagents.html) and (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0034-Ab-binding-proteins.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.