P-PER™ Plant Protein Extraction Reagent - FAQs
P-PER™ Plant Protein Extraction Reagent - FAQs
View additional product information for P-PER™ Plant Protein Extraction Reagent - FAQs (89803)
13 product FAQs found
The P-PER Plant Protein Extraction reagent instructions recommend room temperature storage conditions for P-PER reagents. Would 4 degrees C be better for long-term storage?
For P-PER Reagents A, B and C, room temperature storage is advised. Reagent C tends to become viscous at 4 degrees C. Overall solubility of proteins and higher quality SDS-PAGE analysis are achieved with reagent C storage at room temperature. If the reagents need to be used at 4 degrees C, place Reagents A,B,C at 0-4 degrees C for 1-3 hours before use.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How should I store my extracted proteins after using the P-PER Plant Protein Extraction reagent?
Once you have extracted the sample, do not keep the aqueous phase in contact with the organic phase for more than 30 minutes. If you do not plan to use the proteins immediately within 30 minutes, remove the aqueous phase and store at 4 degrees C or -20 degrees C. Specific applications may require protease inhibitors, phenolic inhibitors and reductants.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What downstream applications is the aqueous buffer compatible with when using the P-PER Plant Protein Extraction reagent?
BCA Reducing Agent Compatible Protein Assay (Cat. No.s 23250, 23252), 1-D gel electrophoresis, 2-D gel electrophoresis (dilute sample 1:10 in sample buffer), Western blot, immunoprecipitation, ion exchange chromatography, polyhistidine-tag protein purification, and activity assays.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Where are the nucleic acids after the P-PER Plant Protein Extraction reagent procedure?
DNA is in the pellet after extraction. You can solubilize the pellet with sample buffer and run this sample on an agarose gel. Without shearing or a restriction digest, you will find genomic (large) DNA. You can use PEI (polyethyleneimine) to precipitate the DNA from any remaining proteins. One of our customers has used these nucleic acids for RT-PCR and was able to isolate RNA. It is recommended to wash the pellet twice with ice-cold Tris buffer and then dissolve in TE buffer.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How can I determine the amount of protein extracted when usign the P-PER Plant Protein Extraction reagent?
The P-PER Kit protein extracts can be quantified using the BCA Protein Assay Kits - Reducing Agent Compatible (Cat. No.s 23250 and 23252).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What if no separation of the phases is observed after Step 6 when using the P-PER Plant Protein Extraction reagent?
Centrifuge the sample for an additional 5 minutes at 2,000 to 5,000 x g. If the tube is dropped after the phase separation, it will not harm the proteins to centrifuge again.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I prepare enough P-PER Working Solution to last for the week?
No. The P-PER Working Solution is prepared just before use. For best results, do not store the Working Solution for more than one day.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What phenolic inhibitor should I use when using the P-PER Plant Protein Extraction reagent?
Phenolics bind to proteins through hydrogen bonds immediately after plant cells are lysed. Thus it is important to use phenolic inhibitors; the two most common being PVPP and PVP. PVPP is the most widely used and is a generic phenolic inhibitor. It binds to phenolics through hydroxyl groups, but can also bind to hydroxyls on proteins resulting in protein loss. It is an insoluble material, like agarose, and takes hours (up to 24) to dissolve into solution. Recommended concentration is 1.5% w/v. PVP is a soluble substance and can be found in a variety of molecular weights. Smaller molecular weights can bind to proteins, so it is recommended to start with 2-4% 40KD PVP. Depending on the specific phenolics in the plant tissue, the molecular weight of PVP will need to be optimized. Different molecular weights bind certain phenolics better than others, so PVP does not bind all phenolics universally.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What protease inhibitor should I use when working with P-PER Plant Protein Extraction reagent?
You can use the same kind of protease inhibitor as is used for mammalian proteins. Although other suppliers sell plant protease inhibitors, there is nothing special about plant samples that require different inhibitors than for mammalian samples. Use EDTA-free Halt Protease Inhibitor (e.g., Cat. No.78437) because EDTA soaks up the calcium. Calcium binds pectin, a sugar that binds proteins; hence, you cannot assay for these proteins. Thus the calcium is necessary to precipitate the pectin so that proteins are free for subsequent analysis. Note: Seeds have high amounts of proteases.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Does the age and storage of the protein source affect protein yield when using the P-PER Plant Protein Extraction reagent?
Yes, as plants age their vacuoles (involved in cell maintenance) get larger. In older plants, 80% of the cell may be comprised of vacuoles. As vacuoles are a mostly liquid compartment, the amount of total protein is decreased as a plant ages. Protein can be extracted from fresh, frozen and dry plant tissue. If the sample has been powdered by freeze/grinding with liquid nitrogen, the extracted protein may no longer be active or useable in functional assays.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is mechanical abrasion necessary with P-PER Plant Protein Extraction reagent?
Yes, you still need mechanical abrasion to disrupt the cell wall of plant cells. The mesh bag in the kit works well for soft plant tissues (e.g., leaves, flowers). You can use a pestle along with the mesh bag to disrupt harder tissues; place the tissue in the bag with the P-PER reagent, then tap the outside of the bag with the pestle.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the ratio of P-PER Plant Protein Extraction reagent to plant tissue?
For fresh/frozen samples, 1ml of aqueous (A+B) + 0.75mL organic (C) + 80mg fresh/frozen plant tissue For dried samples/seeds, 1ml of aqueous (A+B) + 0.75mL organic (C) + 20mg dried plant tissue
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What part of the plant can P-PER Plant Protein Extraction reagent be used for?
Any part: stems, flowers, leaves, seeds, nuts (crack the outer shell and use the nut meat), and tree trunk (will need to grind before P-PER addition).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.