Pierce™ C18 Spin Tips & Columns
Thermo Scientific™

Pierce™ C18 Spin Tips & Columns

Thermo Scientific Pierce C18 Spin Columns (10–150 μL) and Spin Tips (10–100 μL) effectively remove interfering contaminants, while also maintaining excellent binding and recovery of peptide samples for use upstream of mass spectrometry and other high sensitivity detection methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
8987025 ColumnsColumn
8987350 ColumnsColumn
8485096 TipsSpin Tip
8778296 x 10 μL TipsPipette Tip
8778496 x 100 μL TipsPipette Tip
Catalog number 89870
Price (USD)
159.65
Online exclusive
177.00
Save 17.35
Each
Quantity:
25 Columns
Product Type:
Column
Request bulk or custom format
Price (USD)
159.65
Online exclusive
177.00
Save 17.35
Each

C18 Spin Tips for Peptide Desalting and Cleanup

Pierce™ C18 spin tips are designed for rapid, reproducible peptide desalting, cleanup, and concentration prior to mass spectrometry analysis. Utilizing reversed-phase C18 chemistry, these tips help enable efficient peptide binding and recovery while removing salts, detergents, and other contaminants that can interfere with downstream applications.

Used in combination with Pierce Spin Tip Adaptors (Catalog No. A40011776) and low protein binding microcentrifuge tubes (Catalog No. 90410), C18 spin tips support a simple centrifugation-based workflow that delivers consistent and reliable results across a wide range of sample inputs.

Features of C18 Spin Tips

  • Reliable – proven reversed-phase C18 chemistry
  • Efficient – effectively removes interfering contaminants to improve spectral quality
  • Streamlined – simple workflow using tip adaptor and centrifugation steps

Applications

  • Peptide desalting prior to LC-MS analysis
  • Cleanup following protein digestion workflows
  • Removal of salts, buffers, and contaminants
  • Peptide concentration and sample preparation

How to Use

C18 spin tips use a bind–wash–elute workflow performed by centrifugation. Tips are placed into a tip adaptor seated in a microcentrifuge tube for each step. Simply activate and equilibrate prior to adding your acidified sample. To enhance peptide binding repeat the sample loading step one time. Lastly, wash 2-3 times and elute into a fresh tube using an acetonitrile solution. Refer to Table 1 in the Figures for recommended buffers, volumes and spin speeds for each tip size.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypePeptides
For Use With (Equipment)Microcentrifuge, Mass Spectrometer
Quantity25 Columns
SorbentC18
Workflow StepSample Clean Up
Detection MethodMass Spectrometry
Product LinePierce
Product TypeColumn
Starting MaterialPeptides, Protease-digested Protein
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Do I need to do C18 clean-up of my mass spectrometry sample before phosphopeptide enrichment?

Yes. C18 clean-up is necessary to remove small molecules that may interfere with phosphopeptide enrichment. Drying after clean-up is also necessary to concentrate the dilute samples and remove C18 organic eluting solvent so samples can be dissolved in the optimized buffers for phosphopeptide binding. Alternative binding buffers are not recommended as phosphopeptide yield and specificity will be affected. For maximal performance, ensure that the clean, dried peptide samples are completely solubilized in binding buffer with agitation for at least 10 min.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.