Pierce™ Micro-Spin Columns - FAQs
Pierce™ Micro-Spin Columns - FAQs
View additional product information for Pierce™ Micro-Spin Columns - FAQs (89879)
1 product FAQs found
Are Pierce Micro-Spin Columns (Cat. No. 89879) reusable?
Pierce Micro-Spin Columns (Cat. No. 89879) are disposable single-use columns.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.