Sodium Deoxycholate Detergent, 25 g - FAQs
Sodium Deoxycholate Detergent, 25 g - FAQs
View additional product information for Sodium Deoxycholate Detergent - FAQs (89905, 89904)
2 product FAQs found
Are there mass spectrometry reagents for membrane proteins?
CHAPS (Cat. Nos. 28299, 28300), OTG (Cat. No. 28351) and Sodium Deoxycholate (Cat. Nos. 89904, 89905) are detergents often used to solubilize and extract membrane proteins from samples
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What can the sodium deoxycholate detergent be used for?
This detergent is recommended for stripping endotoxin (lps) from immobilized Polymyxin B columns. One could use this product in conjunction with the Thermo Scientific Detoxi-Gel Endotoxin Removing Gel.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.