Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL
Thermo Scientific™

Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL

Perform high-throughput protein recovery (7K MWCO) and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Thermo Scientific Pierce Zeba desalting chromatography columns, plates, and cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatVolume (Metric)
899351 column x 5 mLChromatography Cartridge5 mL
8987725 ColumnsSpin Column75 μL
8987850 ColumnsSpin Column75 μL
A400024695 ColumnsSpin Column20 mL
A4000247510 ColumnsSpin Column20 mL
8989025 ColumnsSpin Column2 mL
898915 ColumnsSpin Column5 mL
8989225 ColumnsSpin Column5 mL
898935 ColumnsSpin Column10 mL
8989425 ColumnsSpin Column10 mL
898895 ColumnsSpin Column2 mL
8988225 ColumnsSpin Column0.5 mL
8988350 ColumnsSpin Column0.5 mL
899345 columns x 1 mLChromatography Cartridge1 mL
898072 Plates96-well Filter Plate96-well
898084 Plates96-well Filter Plate96-well
Catalog number 89935
Price (USD)
262.65
Online exclusive
305.00
Save 42.35
Each
Quantity:
1 column x 5 mL
Format:
Chromatography Cartridge
Volume (Metric):
5 mL
Request bulk or custom format
Price (USD)
262.65
Online exclusive
305.00
Save 42.35
Each
Perform high-throughput protein recovery and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Pierce Zeba desalting chromatography columns, plates, and cartridges. Rapidly desalt protein and biological samples for manual or automated liquid chromatography (LC) with ≥95% retention of salts and other small molecular contaminants (<1,000 Da).

These Zeba spin desalting products contain a proprietary high-performance size-exclusion chromatography resin that enables excellent protein desalting and recovery of 7K molecular weight cutoff (MWCO) molecules using a centrifuge format. This resin is ideal for removing low molecular weight compounds including salts, fluorescent dyes, biotin, and other small labelling reagents.

Benefits of Zeba desalting columns, plates, and cartridges include:

  • High recovery—low-binding resin maximizes protein recovery,
  • Fast—no fraction screening or waiting for protein to emerge by gravity flow
  • Economical—great performance at lower cost than other commercially available cartridges
  • Easy-to-use—no cumbersome column preparation or equilibration
  • Flexible—available in spin columns, filter spin plates, and cartridges for a range of needs 

Zeba Spin Desalting Columns 

Convenient, ready-to-use polypropylene columns are the ideal solution for quick and straightforward desalting and buffer exchange of protein samples in a centrifuge format. The pre-dispensed spin columns are offered in various resin fills (75 μL, 0.5 mL, 2 mL, 5 mL, 10 mL, 20 mL). These options make it simple to handle sample volumes between 2 μL and 10 mL.

Zeba Desalting Plates

Provide trouble-free high-throughput desalting and buffer exchange for sample volumes ranging from 20 to 100 μL. Pre-dispensed 96-well filter plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in five minutes. Compatible with centrifugation for manual or automated purification.

Zeba Desalting Chromatography Cartridges

Compatible with automated liquid chromatography systems and with syringe processing, Zeba desalting cartridges provide fast, easy, and reproducible chromatographic separations and can be regenerated for multiple uses.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeDesalting Chromatography Cartridge
Purification TargetBuffer Exchange, Protein
Quantity1 column x 5 mL
Volume (Metric)5 mL
Column TypeSize-exclusion, Proprietary Resin
FormatChromatography Cartridge
MWCO7.0 kDa
Product LineZeba
Sample Volume (Metric)0.1 to 1.5 mL
Unit SizeEach
Contents & Storage
Upon receipt store at 4–8°C.

Frequently asked questions (FAQs)

Can Zeba Desalting columns, plates, and cartridges be autoclaved?

Yes, Zeba Desalting columns, plates, and cartridges can be autoclaved at 115 degrees C for 30 min.

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?

Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

Can Zeba Desalting Chromatography Cartridges be regenerated and stored after use?

Yes, Zeba Desalting Chromatography Cartridges can be regenerated and stored for later use. The catridges may be reused multiple times without significant loss of performance.

For regeneration, wash the resin with 3-5 column volumes of water or buffer before applying the next sample. For storage, wash the resin with 5 column volumes of 0.02-0.05% sodium azide in water. Attach the supplied bottom cap and then the top plug and store the cartridge at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

How should I store Zeba Spin Desalting Columns, Plates, and Cartridges?

We recommend storing Zeba Spin Desalting Columns, Plates, and Cartridges at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

What product do you recommend for removing sodium azide from the Alix Monoclonal Antibody (3A9) (Cat. No. MA1-83977) storage solution?

We recommend using Zeba Spin Desalting Columns, Plates, and Cartridges (Cat. No. 89882, 89883) to remove sodium azide from the Alix Monoclonal Antibody (3A9) storage solution.