EDTA disodium salt dihydrate, 99.01-101.0% (dried basis), crystals, USP, GMP, J.T.Baker™
Avantor™

EDTA disodium salt dihydrate, 99.01-101.0% (dried basis), crystals, USP, GMP, J.T.Baker™

GMP Manufactured Product. Meets U.S.P Requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8994-0712 kg
8994-01500 g
Catalog number 8994-07
Price (USD)
-
Quantity:
12 kg

GMP Manufactured Product. Meets U.S.P Requirements.

Specifications
CAS6381-92-6
Chemical Name or MaterialEDTA Disodium Salt Dihydrate
ColorWhite
Melting Point252°C
Molecular FormulaC10H14N2Na2O8·2H2O
Packaging TypePolyethylene pail, screw top
Purity99.01 to 101.0%
Quantity12 kg
SynonymEthylenedinitrilotetra-acetic acid disodium salt dihydrate
GradeUSP
Unit SizeEach