HisPur™ Cobalt Spin Columns, 0.2 mL
Thermo Scientific™

HisPur™ Cobalt Spin Columns, 0.2 mL

Thermo Scientific HisPur Cobalt Spin Columns contain a tetradentate chelating agarose resin charged with divalent cobalt (Co2+) for obtaining high-purityRead more
Have Questions?
Catalog NumberQuantity
8996725 Columns
Catalog number 89967
Price (USD)
291.00
Each
Quantity:
25 Columns
Request bulk or custom format
Price (USD)
291.00
Each
Thermo Scientific HisPur Cobalt Spin Columns contain a tetradentate chelating agarose resin charged with divalent cobalt (Co2+) for obtaining high-purity his-tagged proteins with no metal contamination.

Features of HisPur Cobalt Resin:

• High purity—obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
• Specificity—cobalt-chelate coordination core binds fewer host protein contaminants, resulting in lower background than nickel resins
• Low metal leaching—no metal contamination in eluted histidine-tagged protein sample
• Versatility—purify proteins under native or denaturing conditions; compatible with Thermo Scientific Pierce Cell Lysis Reagents and a variety of buffer additives
• Cost effective—resin can be reused several times or discarded after single use
• Flexibility—available as bulk resin, kits, predispensed columns, chromatography cartridges and 96-well filter plates

Compared to Ni-IDA and tetradentate Ni2+ chelate resins, HisPur Cobalt Resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole). Although Ni(2+) chelate resins achieve high protein yields, they bind somewhat indiscriminantly, resulting in suboptimal purity. Often, additional cleanup steps are required. By contrast, cobalt maximizes protein purity without sacrificing protein yield. HisPur Cobalt Resin binds fewer nonspecific proteins, displays less metal leaching and enables less stringent elution conditions than nickel resins.

HisPur Cobalt IMAC Resin specifically binds His-tagged recombinant protein that can be efficiently recovered by mild imidazole elution. Greater than 10 mg of His-tagged protein can be bound per milliliter of resin using a 28 kDa fusion protein expressed and purified from an E. coli lysate.

Various His-tagged proteins can be recovered in similar or higher purity compared to that obtained from other available nickel or cobalt IMAC resins using either a spin or gravity-flow column format. Purification performance of HisPur Cobalt Resin was benchmarked against IMAC resins from other suppliers using multiple His-tagged proteins of different sizes and expression levels. The results demonstrate that HisPur Cobalt Resin achieves maximal protein purity without sacrificing protein yield.

Available Formats:
• Resin Slurries—crosslinked 6% beaded agarose; 10 mL, 100 mL, 500 mL bottles
• Spin Columns—0.2 mL (microcentrifuge), 1 mL, and 3 mL columns
• Purification Kits—complete kits in all three column sizes
• Chromatography Cartridges—learn more and see HisPur Chromatography Cartridge data
• 96-well Spin Plates

Related Products
HisPur™ Cobalt Resin
HisPur™ Cobalt Purification Kit, 0.2 mL
HisPur™ Cobalt Spin Plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)0.2 mL
Column TypeAffinity, Agarose Resin
DescriptionHisPur Cobalt Spin Columns, 0.2mL
FormatMicrocentrifuge, Spin Column
Protein FormRecombinant
Quantity25 Columns
Stationary PhaseMetal Chelate, Divalent Cobalt
TargetHis-Tagged Protein
FormLiquid Suspension
Product LineHisPur
TypeSpin Column
Unit SizeEach
Contents & Storage
Store at 4°C

Frequently asked questions (FAQs)

What is the difference between HisPur Nickel Resin and HisPur Cobalt Resin?

Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.

HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.

HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.