GeneChip™ DNA Labeling Reagent, 7.5 mM - FAQs
GeneChip™ DNA Labeling Reagent, 7.5 mM - FAQs
View additional product information for GeneChip™ DNA Labeling Reagent, 7.5 mM - FAQs (900542)
1 product FAQs found
What is the shelf life of this the DNA Labeling reagent (Cat. No. 900542)?
The DNA Labeling Reagent has about 12 months of dating from the date of shipment.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.