TMTsixplex™ Isobaric Label Reagent Set
Thermo Scientific™

TMTsixplex™ Isobaric Label Reagent Set

Amine-reactive Thermo Scientific™ TMT Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues from up to 6 samples using tandem mass spectrometry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
900622 x 0.8 mg (per tag)
900611 x 0.8 mg (per tag)
900665 x 0.8 mg (per tag)
900682 x 5 mg (per tag)
Catalog number 90062
Price (USD)
1,337.65
Online exclusive
1,408.00
Save 70.35
Each
Quantity:
2 x 0.8 mg (per tag)
Price (USD)
1,337.65
Online exclusive
1,408.00
Save 70.35
Each

Tandem Mass Tag™ 6-plex (TMTsixplex™) Reagents are sets of isobaric compounds (i.e., same mass and structure, also called isotopomers) that are NHS-activated for covalent, irreversible labeling of primary amines (–NH2) groups. Each isobaric reagent contains a different number of heavy isotopes in the mass reporter region, which results in a unique reporter mass during tandem MS/MS for sample identification and relative quantitation. Amine-reactive Thermo Scientific™ TMT Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues from up to 6 samples using tandem mass spectrometry.

Features of TMTsixplex Label Reagents and Kits:

  • Powerful – multiplexed MS analysis increases sample throughput and enables relative quantitation of up to six different samples derived from cells, tissues or biological fluids
  • Robust – consistent chemistry allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research
  • Efficient – amine-reactive NHS-ester activated reagents ensures greater than 95% labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Compatible – optimized for use with high resolution Thermo Scientific™ Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer™ 2.3 and above

Applications of TMTsixplex Label Reagents and Kits:

  • Protein identification and quantitation from multiple samples of cells, tissue or biological fluids
  • Protein expression profiling of normal vs. disease states or control vs. treated
  • Quantitative analysis of proteins for which no antibodies are available
  • Identification and quantitation of membrane and post-translationally modified proteins
  • Identification and quantitation of hundreds to thousands of proteins in a single experiment

Since TMT reagents share an identical structure and mass (i.e., isotopomers), labeled peptides co-elute during LC separation and are co-isolated during MS/MS analysis, resulting in fewer missing peptide identifications among samples. During MS/MS analysis, each isobaric tag is also fragmented to produce a unique reporter ion mass that is used for sample identification and quantitation. Protein quantitation is accomplished by comparing the relative intensities of the six reporter ions.

TMT Reagents are provided as standalone sets or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. Tandem Mass Tag Reagents combined with high resolution Thermo Scientific™ Orbitrap instruments and Proteome Discoverer software provide integrated total system solutions for quantitative protein expression analysis. For high resolution analysis of the TMT-labeled peptides, the recommended LC column for Nanospray Flex source is Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For EASY-Spray source, the recommended LC column is EASY-Spray C18 LC Column (Cat. No. ES902 or ES903).

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Thompson, A., et al.(2003). Tandem mass tags: a novel quantification strategy for comparative analysis of complex protein mixtures by MS/MS. Anal. Chem.75(8), 1895-1904.
  2. Dayon, L., et al.(2008). Relative quantification of proteins in human cerebrospinal fluids by MS/MS using 6-plex isobaric tags. Anal. Chem. 80(8), 2921 -2931.
Specifications
DescriptionTMTsixplex™ Isobaric Label Reagent Set, 2 x 0.8 mg
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeTandem Mass Tag™ (TMT) Labels
Quantity2 x 0.8 mg (per tag)
Workflow StepProtein Labeling
Detection MethodMass Spectrometry
Product LineTMTsixplex
Starting MaterialCell Lysate
Unit SizeEach
Contents & Storage
Sufficient for: Two sixplex (2 x 6-way) experiments
• TMT6-126 Label Reagent, 2 x 0.8 mg
• TMT6-127 Label Reagent, 2 x 0.8 mg
• TMT6-128 Label Reagent, 2 x 0.8 mg
• TMT6-129 Label Reagent, 2 x 0.8 mg
• TMT6-130 Label Reagent, 2 x 0.8 mg
• TMT6-131 Label Reagent, 2 x 0.8 mg

Store at –20°C.

Frequently asked questions (FAQs)

What type of mass spectrometer can I use for Tandem Mass Tag (TMT) analysis?

We recommend using high-resolution Orbitrap Tribrid (e.g., Fusion, Lumos, Eclipse), Orbitrap Exploris (e.g., 240, 480), or Q Exactive (e.g,. Plus, HF, HFX) series of instruments.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

With the TMTsixplex Isobaric Label Reagent Set (Cat. Nos. 90061, 90062), what is the smallest amount of protein that can be labeled?

The smallest amount of protein that can be labeled is 10 ug, and the volumes for reduction, alkylation, and digestion need to be adjusted. In order to get a good labeling efficiency, we recommend using a peptide: TMT ratio of 1:8.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is TMT quantitation relative or absolute?

TMT quantitation is relative between samples unless a heavy peptide standard is included for comparison.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the main purpose of TMT (Tandem Mass Tag) reagents?

TMT technology is used to perform multiplex quantitation of proteins extracted from cells and tissues. Rather than quantify single proteins (for example with an antibody), here, large numbers of proteins can be quantified in a single run.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.