GeneChip™ S. cerevisiae Tiling 1.0R Array - FAQs
GeneChip™ S. cerevisiae Tiling 1.0R Array - FAQs
View additional product information for GeneChip™ S. cerevisiae Tiling 1.0R Array - FAQs (900645)
2 product FAQs found
Do you offer any annotation files for the GeneChip S. cerevisiae Tiling 1.0R Array?
We do not provide annotation files for the GeneChip S. cerevisiae Tiling 1.0R Array, but we do provide annotation files for human and mouse promoter arrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Is the GeneChip S.cerevisiae Tiling 1.0R Array designed from the forward strand or the reverse strand?
In the name of this product, GeneChip S. cerevisiae Tiling 1.0R Array, the 'R' indicates that the probe was designed from the reverse strand.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.