B-PER™ with Enzymes Bacterial Protein Extraction Kit
Thermo Scientific™

B-PER™ with Enzymes Bacterial Protein Extraction Kit

Thermo Scientific B-PER with Enzymes Bacterial Protein Extraction Kit is an easy-to-use cell lysis reagent is a nonionic detergent-based solutionsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
90079500 mL
90078250 mL
Catalog number 90079
Price (USD)
753.65
Online exclusive
828.00
Save 74.35
Each
Quantity:
500 mL
Price (USD)
753.65
Online exclusive
828.00
Save 74.35
Each
Thermo Scientific B-PER with Enzymes Bacterial Protein Extraction Kit is an easy-to-use cell lysis reagent is a nonionic detergent-based solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. All B-PER Reagents are compatible with downstream applications, such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay). Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Features of the B-PER Bacterial Protein Extraction Reagent:

• Tris buffered formulation with lysozyme and DNase enzymes
• Improved cell membrane solubilization and DNA digestion

All Thermo Scientific B-PER Protein Extraction Reagents are:
• Ready to use—one-step cell lysis of gram-positive and gram-negative bacteria using a mild, nonionic detergent (proprietary) in Tris or phosphate buffer formulations
• Fast and simple—just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
• Convenient – B-PER Complete Reagent contains lysozyme and a universal nuclease in a single formulation with 4°C storage
• Excellent yields—recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
• Flexible—B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
• Compatible—completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications

Convenient, Ready-to-use Formats
B-PER Bacterial Extraction Reagents are more effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications. B-PER Reagents are formulated in Tris or phosphate buffer at physiological pH. They extract native and soluble recombinant proteins and yield lysates that are directly compatible with most downstream workflows such as electrophoresis, affinity purification, immunoprecipitation, protein interaction analysis, crosslinking and protein labeling.

More Product Data
• Improved, all-in-one B-PER Reagent for bacterial protein extraction

Related Products
B-PER™ Bacterial Protein Extraction Reagent
B-PER™ Complete Bacterial Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
Quantity500 mL
Volume (Metric)500 mL
Product LineB-PER
Product TypeBacterial Protein Extraction Kit
Unit SizeEach
Contents & Storage
Sufficient For: 125g cell paste
• B-PER Reagent, 500 mL
• Lysozyme, 50 mg/mL, 2 x 0.5 mL
• DNase I, 2500 U/mL, 2 x 0.5 mL

Store B-PER Reagent at ambient temperature. Store Lysozyme and DNaseI at -20°C.

Frequently asked questions (FAQs)

What is the difference between B-PER and B-PER II Reagent?

B-PER II Reagent contains more detergent making it ideal for extracting proteins from small bacterial cultures with less than 20 mL in volume.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the composition of B-PER Reagent?

B-PER Reagent utilizes a proprietary, mild nonionic detergent in a 20 mM Tris HCl, pH 7.5 buffer. No enzymatic components are present in the B-PER solution itself (B-PER Complete Bacterial Protein Extraction Reagent, Cat. No. 89821, 89822 does contain enzymes). Depending on your particular protein, you may need to add components such as salt, lysozyme, protease inhibitors, reducing agents and chelating agents. Other ready-to-use formats include B-PER II Bacterial Protein Extraction Reagent (2X), B-PER Reagent (in Phosphate Buffer), and B-PER with Enzymes Bacterial Protein Extraction Kit.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the volume of B-PER Extraction Reagent to use per gram weight of wet cells?

For 1 gram of cells add 4 mL of B-PER Reagent.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Will my protein be in its native conformation after B-PER Reagent extraction?

While this is protein-dependent, many proteins have been successfully tested in downstream applications (reporter assays, immunoprecipitation and beta-Gal assays), including GST and 6xHis proteins. Otherwise, samples can be diluted or dialyzed to remove any interfering substances.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Why is my protein extract so viscous after B-PER Reagent extraction?

This is often an indication of the presence of large amounts of DNA in the extract. This viscosity will be greatly decreased upon addition of DNase I (Cat. No. 90083).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.