iodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg (per tag) - FAQs
iodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg (per tag) - FAQs
View additional product information for Thermo Scientific™ iodoTMT Isobaric Label Reagents and Kits - FAQs (90101, 90100, 90102)
1 product FAQs found
What is the difference between IodoTMT tags and Iodoacetamido-LC-Phosphonic Acid?
Both IodoTMT tags and Iodoacetamido-LC-Phosphonic Acid react with sulfhydryl groups. Iodo TMT tags allow the quantitation of proteins with cysteine amino acids and the oxidative state of sulfhydryl groups, while Iodoacetamido-LC-Phosphonic Acid allow the detection and enrichment of those specific proteins. This is a great tool for mass spectrometry (MS) analysis, and High-Select Fe-NTA Magnetic Phosphopeptide Enrichment Kit (Cat. No. A52283), can be used for the enrichment step.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.