Does Ref103 come in the CytoScan XON Assay Kit (Cat. No. 931311)?
Reference Genomic DNA 103 is not included in the kit, but is available for purchase separately (Cat. No. 901012).
Note: A positive control is recommended for the CytoScan XON assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Note: A positive control is recommended for the CytoScan XON assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.