Axiom™ Biobank Genotyping Array, 96 array plate - FAQs
Axiom™ Biobank Genotyping Array, 96 array plate - FAQs
View additional product information for Axiom™ Biobank Genotyping Array, 96 array plate - FAQs (902186)
4 product FAQs found
For the Axiom Biobank Genotyping Array, 96 array plate (Cat. No. 902186), which instrument is required to run the assay?
The Beckman Biomek™ FXP Target Prep Express System is required for running the Axiom Biobank Genotyping Array, 96 array plate (Cat. No. 902186).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.