GeneChip™ Human Transcriptome Assay 2.0, 30 reactions - FAQs
GeneChip™ Human Transcriptome Assay 2.0, 30 reactions - FAQs
View additional product information for GeneChip™ Human Transcriptome Assay 2.0 - FAQs (902309, 902310)
2 product FAQs found
How long does it take for the GeneChip Human Transcriptome Array 2.0 (Cat. No. 902162) to scan?
The GeneChip Human Transcriptome Assay 2.0 takes approximately 35 min to scan.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.