Product Image
Applied Biosystems™
RNA Quantification Kit for SYBR Green I and ROX™ Passive Reference Dye
Target preparation assay kits such as GeneChip™ WT Pico Kit and GeneChip 3' IVT Pico Kit allow expression profiling using as little as 100 pg of total RNA (approximately 10 cells).
Have Questions?
Catalog NumberQuantity
9029051 kit
Catalog number 902905
Price (USD)
343.00
Each
Quantity:
1 kit
Price (USD)
343.00
Each
Accurate quantification from limited RNA concentrations
Target preparation assay kits such as GeneChip™ WT Pico Kit and GeneChip 3' IVT Pico Kit allow expression profiling using as little as 100 pg of total RNA (approximately 10 cells). These low amounts of RNA for such target preparation assays require accurate measurement of the quantity of input sample RNA for optimal performance. However, at these low RNA amounts, it becomes virtually impossible to accurately quantify the RNA with standard UV absorbance or fluorescent dye–based techniques.
The RNA quantification kit uses Power SYBR Green RNA-to-CT 1-Step Kit to measure levels of the consistently expressed 18S ribosomal RNA (rRNA) in human, mouse, and rat samples. The amount of 18S rRNA target in the unknown samples is then scaled against a Control HeLa RNA standard curve for accurate quantification that fails using conventional techniques. In addition to the Power SYBR Green RNA-to-CT 1-Step Kit, the kit also includes Control HeLa Total RNA for standard curve preparation and an 18S rRNA Primer Mix, providing an assay range of 0.01 pg–1 ng total RNA.
*Check real-time PCR instrument for selecting appropriate passive reference dye.
Related Links:
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)RNA
Product TypeRNA Quantification Kit
Quantitation Range0.01 pg–1 ng total RNA
Quantity1 kit
FormatKit
Unit SizeEach
Frequently asked questions (FAQs)
Can I use FFPE samples with the RNA Quantification Kit for SYBR Green I and ROX Passive Reference Dye?
No, we do not recommend using FFPE samples with the Applied Biosystems RNA Quantification Kit. The purpose of the RNA Quantification Kit is to determine if a degraded or low input sample has enough total RNA to run a WT and/or IVT Pico assay. If this kit is used with FFPE samples, the amount of RNA in the sample may be overestimated due to interference from reagents used when formalin-fixing and paraffin embedding the samples.