Search
View additional product information for PharmacoScan™ Assay Kit, 24-Format - FAQs (903010TS)
1 product FAQs found
We recommend reviewing the Microarrays for Preemptive Pharmacogenomics page to determine a potential alternative.
If you have any questions you can email Tech Support at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.