Pierce™ MS-Compatible Magnetic IP Kits
Thermo Scientific™

Pierce™ MS-Compatible Magnetic IP Kits

Achieve highly efficient, compatible immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using Thermo Scientific Pierce MS-Compatible Magnetic IP Kits with streptavidin or Protein A/G magnetic beads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
90409Protein A/G Magnetic Beads
90408Streptavidin Magnetic Beads
Catalog number 90409
Price (USD)
597.65
Online exclusive
672.00
Save 74.35
Each
Description:
Protein A/G Magnetic Beads
Request bulk or custom format
Price (USD)
597.65
Online exclusive
672.00
Save 74.35
Each
Immunoprecipitation is often necessary for the detection and quantitation of low-abundance proteins or rare signaling events. Mass spectrometry analysis relies on two methods for the detection of low-abundance proteins: single reaction monitoring (SRM) and parallel reaction monitoring (PRM), which enable isolation, scheduling, and quantitation of selected proteotypic peptides. These IP kits have been optimized for efficient immunoprecipitation and co-immunoprecipitation of target proteins, and are directly compatible with downstream MS sample preparation and analysis.
• MS-compatible—reagents directly compatible with in-solution peptide digestion
• Sensitive—procedure successfully enriches low-abundance proteins (low ng range)
• Low background—enriched samples contain minimal detergent residuals
• Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods
• Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein

The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.

Applications
• Enrichment of low-abundance targets to improve MS sensitivity
• Identification of target proteins that are not detectable by western blotting
• Quantitation of target proteins by LC-MS analysis
• Screening of antibodies for IP-MS

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionProtein A/G Magnetic Beads
Final Product TypeProtein
For Use With (Application)Mass Spectrometry
For Use With (Equipment)KingFisher™ Flex, Magnetic Stand
Quantity40 reactions kit
Sufficient For40 Reactions
Workflow StepSample Enrichment
Detection MethodMass Spectrometry
FormatKit
Product LinePierce
Product TypeMagnetic IP Kit
Starting MaterialProtein Samples, Cell Lysate
Unit SizeEach
Contents & Storage
Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP-MS Elution Buffer, 6 mL
MgCl2 (1 M), 0.75 mL

Store in refrigerator (2° to 8°C).

Frequently asked questions (FAQs)

What is the difference between SureQuant IP-MS Sample Preparation kits and Pierce MS-Compatible Magnetic IP kits?

SureQuant IP-MS Sample Preparation kits include all reagents necessary for sample lysis, immunoenrichment, elution, digestion, and MS analysis. The entire process is completed in 4 hr for same-day LC-MS/MS analysis as compared to traditional IP methods that usually take around 21 hr. Pierce MS-Compatible Magnetic IP kits provide reagents necessary for the lysis step and the elution step, but not for the digestion step prior to MS analysis.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.