Pierce™ MS-Compatible Magnetic IP Kit, Protein A/G, 40 reactions kit - FAQs
Pierce™ MS-Compatible Magnetic IP Kit, Protein A/G, 40 reactions kit - FAQs
View additional product information for Pierce™ MS-Compatible Magnetic IP Kits - FAQs (90408, 90409)
1 product FAQs found
What is the difference between SureQuant IP-MS Sample Preparation kits and Pierce MS-Compatible Magnetic IP kits?
SureQuant IP-MS Sample Preparation kits include all reagents necessary for sample lysis, immunoenrichment, elution, digestion, and MS analysis. The entire process is completed in 4 hr for same-day LC-MS/MS analysis as compared to traditional IP methods that usually take around 21 hr. Pierce MS-Compatible Magnetic IP kits provide reagents necessary for the lysis step and the elution step, but not for the digestion step prior to MS analysis.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.