Low Protein Binding Collection Tubes (1.5 mL) - FAQs
Low Protein Binding Collection Tubes (1.5 mL) - FAQs
View additional product information for Low Protein Binding Microcentrifuge Tubes - FAQs (90411, 88380, 90410, 88379)
1 product FAQs found
Can Low Protein Binding Microcentrifuge Tubes be autoclaved?
The Low Protein Binding Collection Tubes are made of polypropylene, which is generally resistant to autoclaving. Therefore, they should be fine to autoclave, but we cannot provide a guarantee.