PharmacoScan™ Training Kit, 96-format
Applied Biosystems™

PharmacoScan™ Training Kit, 96-format

The Applied Biosystems PharmacoScan Training Kit includes two 96-format array plates plus reagents, controls, and consumables to process 94 samples and 2 controls.
Have Questions?
Catalog NumberQuantity
91302794 samples
Catalog number 913027
Price (USD)
-
Quantity:
94 samples

The Applied Biosystems PharmacoScan Training Kit includes two 96-format array plates plus reagents, controls, and consumables to process 94 samples and 2 controls. Two DNA training plates containing 94 known samples each are also included.

Note: This kit includes all consumables and reagents found in the PharmacoScan Assay Kit and enough material to process two 96-format array plates. Two training plates containing known DNA controls are also included.

PharmacoScan Solution is an ideal platform for translational researchers in healthcare, pharma, and research organizations looking to understand how genetic variations affect drug absorption, distribution, metabolism, and excretion (ADME). By pre-emptively screening for risk factors on a microarray, users in research can precisely and reproducibly analyze 4,627 high- to low-evidence markers* within nearly 1,200 genes* in a single assay. The solution includes copy number variation calling and interrogating predictive markers in complex genes. Unlike solutions that analyze only high-evidence markers, PharmacoScan Solution presents a more comprehensive view for genotyping for pharmacogenomic risk.

*Disclaimer: Number of variants genotyped by default as well as variant to gene mapping is refined over time.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Microarray Analysis
Product LinePharmacoScan
Quantity94 samples
TypeTraining Kit
ArrayGenotyping
Format96-array Plate
Number of Arrays2 arrays
SpeciesHuman
Unit SizeEach
Contents & Storage
• 2 x 96-format array plates; store at 2–8°C
• 2 training plates containing known DNA samples; store at -5 to -30°C.
• 1 reagent kit (includes reagents and controls for 2 x 96-format array plates). Storage can vary for each reagent; refer to product insert.
• Gene Titan instrument consumables for 2 x 96-format array plates; store at room temp.