96-Well Microtiter™ Microplates, Polystyrene, 330 μL, Nonsterile, Flat-bottom - FAQs
96-Well Microtiter™ Microplates, Polystyrene, 330 μL, Nonsterile, Flat-bottom - FAQs
View additional product information for 96-Well Microtiter™ Microplates - FAQs (2205, 9205, 2305-11, 2605)
1 product FAQs found
What is the well shape and well capacity for 96-Well Microtiter Microplates (Cat. No. 9205)? Do they have a lid?
96-Well Microtiter Microplates (Cat. No. 9205) do not have a lid and have the following well shape: curved edges, flat bottom (C-well). Each well can hold 330 µL volume.