3UV Lamp (115V, 60Hz) - FAQs
3UV Lamp (115V, 60Hz) - FAQs
View additional product information for 3UV Lamp (115V, 60Hz) - FAQs (95034)
1 product FAQs found
Do you offer replacement bulbs for 3UV Lamp (115V, 60Hz) (Cat. No. 95034)?
The catalog numbers for replacement bulbs for 3UV Lamp (115V, 60Hz) are as follows:
  • Short wave: 34-0013-01
  • Long wave 34-0034-01
  • Mid-range 34-0044-01