Clariom™ GO Screen Assay, human - FAQs
Clariom™ GO Screen Assay, human - FAQs
View additional product information for Clariom™ GO Screen Assay, human - FAQs (952361)
7 product FAQs found
Where can I find the library files for the Clariom GO Screen Assay?
See the Support Files section on the Clariom GO Screen Assay product page (https://www.thermofisher.com/order/catalog/product/952361) for related documents and library files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What type of data analysis do you provide for the Clariom GO Screen Assay?
Basic data analysis is provided by our service providers and includes QC and data normalization, statistical tests for differential expression, and analysis of genes and pathways. Our free and easy-to-use Transcriptome Analysis (TAC) software is also available for download from the website (https://www.thermofisher.com/us/en/home/life-science/microarray-analysis/microarray-analysis-instruments-software-services/microarray-analysis-software/affymetrix-transcriptome-analysis-console-software.html) if you wish to conduct analysis of the data.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Who are the service providers that can run the Clariom GO Screen Assay for me?
We have service providers in the United States and Europe, verified to run the Clariom GO Screen Assay. Please contact your local representative to learn more about our service providers and to discuss shipping options.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When should I use Cells-to-Signal lysis vs. purified RNA for the Clariom GO Screen Assay?
Generally, the more samples you are running for the Clariom GO Screen Assay, the more time and cost will be saved with direct Cells-to-Signal lysis.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which cell lines can I lyse with the Cells-to-Signal Kit for use with the Clariom GO Screen Assay?
Two cell lines have been fully verified for the Clariom GO Screen assay, HepG2 and HeLa. General guidance for creating lysates with other cell lines is included in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018542_ClariomGO_Screen384HT_UG.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How much RNA do I need to run the Clariom GO Screen Assay?
We recommend using at least 500 pg of RNA, but the range of the assay is 100 pg - 10 ng RNA input. See the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018542_ClariomGO_Screen384HT_UG.pdf) for further details.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I order the Clariom GO Screen Assay?
You can purchase the Clariom GO Screen Assay (Cat. No. 952361). To start a GO Screen project, contact your local representative and/or complete the quote request form via the product page (https://www.thermofisher.com/order/catalog/product/952361). Our verified service providers are available to help you run your GO Screen assay and can assist with the management of your project.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.